missing translation for 'onlineSavingsMsg'
Learn More

17Beta-Dihydro Equilenin, TRC

Product Code. 30010789
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010789 10 mg 10mg
30011611 2.5 mg 2.5mg
30007495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010789 Supplier Toronto Research Chemicals Supplier No. TRCD44896010MG
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1423-97-8
Molecular Formula C18 H20 O2
Molecular Weight (g/mol) 268.35
Synonym Estra-1,3,5,7,9-pentaene-3,17-diol, (17β)-, Estra-1,3,5(10),6,8-pentaene-3,17β-diol (6CI,7CI), Estra-1,3,5,7,9-pentaene-3,17β-diol (8CI), (17β)-Estra-1,3,5,7,9-pentaene-3,17-diol, 17β-Dihydroequilenin, E 380, β-Dihydroequilenin
IUPAC Name (13S,14S,17S)-13-methyl-11,12,14,15,16,17-hexahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CCc3c(ccc4cc(O)ccc34)[C@@H]1CC[C@@H]2O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16-17,19-20H,6-9H2,1H3/t16-,17-,18-/m0/s1
Formula Weight 268.1463
Percent Purity >95%
Chemical Name or Material 17beta-Dihydroequilenin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.