missing translation for 'onlineSavingsMsg'
Learn More

17Beta-Dihydro Equilin, TRC

Product Code. 30009024
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009024 1 mg 1mg
30007441 25 mg 25mg
30006944 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009024 Supplier Toronto Research Chemicals Supplier No. TRCD4491301MG
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3563-27-7
Molecular Formula C18 H22 O2
Molecular Weight (g/mol) 270.37
Synonym Estra-1,3,5(10),7-tetraene-3,17-diol, (17β)-, Estra-1,3,5(10),7-tetraene-3,17β-diol (6CI,7CI,8CI), (17β)-Estra-1,3,5(10),7-tetraene-3,17-diol, 17β-Dihydroequilin, 7-Dehydroestradiol, NSC 12170, β-Dihydroequilin
IUPAC Name (9S,13S,14S,17S)-13-methyl-6,9,11,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3C(=CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16-17,19-20H,2,6-9H2,1H3/t14-,16+,17+,18+/m1/s1
Formula Weight 270.162
Chemical Name or Material 17beta-Dihydroequilin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.