missing translation for 'onlineSavingsMsg'
Learn More

17beta-Estradiol, TRC

Product Code. 30041189
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041189 5 g 5g
30041172 1 g 1g
17969929 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041189 Supplier Toronto Research Chemicals Supplier No. TRCE8880005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18 H24 O2
Molecular Weight (g/mol) 272.38
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, Estradiol (8CI), (17β)-Estra-1,3,5(10)-triene-3,17-diol, (+)-3,17β-Estradiol, 13β-Methyl-1,3,5(10)-gonatriene-3,17β-ol, 17β-Estradiol, 17β-Oestradiol, 3,17-Epidihydroxyestratriene, 3,17β-Dihydroxyestra-1,3,5(10)-triene, 3,17β-Estradiol, Absorlent Matrix, Aerodiol, Alora, Altrad, Aquadiol, Bardiol, Benzogynestry, Beta-estradiol, Bio-E Gel, Climaderm, Climara, Compudose, Compudose 100, Compudose 200, Compudose 365, Compudose 400, Corpagen, Dermestril, Destradiol, Dihydrofollicular hormone, Dihydrofolliculin, Dihydromenformon, Dihydrotheelin, Dihydroxyestrin, Dimenformon, Diogyn, Diogynets, Divigel, E 2, Elestrim, Encore, Epiestriol 50, Estasorb, Estra-1,3,5(10)-triene-3,17-diol, (17β)-, Estra-1,3,5(10)-triene-3,17β-diol, Estrace, Estraderm, Estraderm MX, Estraderm TTS, Estraderm TTS 100, Estraderm TTS 50, Estradiol-17β, Estradot, Estraldine, Estrapatch, Estredox, Estreva, Estring, Estring Vaginal Ring, Estroclim, Estroclim 50, Estrogel, Estrogel HBF, Estropause, Estrovite, Evamist, Evorel, FemSeven, Femanest, Femestral, Femogen, Fempatch, Follicyclin, Gelestra, Ginosedol, Gynergon, Gynodiol, Gynoestryl, Innofem, Lamdiol, Macrodiol, Menorest, Menostar, NSC 9895, Nordicol, Oesclim, Oestergon, Oestra-1,3,5(10)-triene-3,17β-diol, Oestradiol, Oestrogel, Oestroglandol, Ovahormon, Ovasterol, Ovastevol, Ovocyclin, Ovocylin, Perlatanol, Primofol, Profoliol, Profoliol B, Progynon, Progynon DH, Sandrena 1, Sprediol, Syndiol, Systen, Theelin, dihydro-, Vagifem, Vivelle, Vivelle-Dot, Zesteem, Zumenon, β-Estradiol
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1
Formula Weight 272.1776
Percent Purity >95%
Chemical Name or Material Estradiol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.