missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Propeller Stirrer

Product Code. 10080791
Click to view available options
Length (Metric):
500 mmH
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10080791

Brand: Bochem™ 9111

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Contains three fix blades.

  • Rpm-range middle
  • Mixing of media with low and middle viscosity
  • Ideal for homogenizing and suspensioning
  • Axial flow

Specifications

Diameter (Metric) 70 mm
Diameter (Metric) Shaft 8 mm
For Use With (Application) Homogenizing and Suspensioning
Length (Metric) 500 mmH
Quantity 1 Each
Speed Medium to High
Type Propeller Stirrer
Viscosity Rating Low to Medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.