missing translation for 'onlineSavingsMsg'
Learn More

18-Crown-6, ≥99.0% (GC), Honeywell™ Fluka™

Product Code. 15640870 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15640870 5 g Glass Bottle 5g
15620870 10 g Poly Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15640870 Supplier Honeywell-Fluka Supplier No. 076735G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17455-13-9
Molecular Formula C12H24O6
Molecular Weight (g/mol) 264.32
MDL Number MFCD00005113
InChI Key XEZNGIUYQVAUSS-UHFFFAOYSA-N
Synonym 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane
PubChem CID 28557
ChEBI CHEBI:32397
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCO1

Specifications

Melting Point 42°C to 45°C
Color Colorless
Flash Point >113°C (>235.4°F)
Packaging Glass Bottle
Quantity 5 g
UN Number NONH for all modes of transport
Beilstein 1619616
Formula Weight 264.32g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material 18-Crown-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.