missing translation for 'onlineSavingsMsg'
Learn More

18-Crown-6, ≥99.0% (GC), Honeywell™ Fluka™

Product Code. p-7120023 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 15640870

Brand: Honeywell-Fluka 076735G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17455-13-9
Molecular Formula C12H24O6
Molecular Weight (g/mol) 264.32
MDL Number MFCD00005113
InChI Key XEZNGIUYQVAUSS-UHFFFAOYSA-N
Synonym 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane
PubChem CID 28557
ChEBI CHEBI:32397
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCO1

Specifications

Melting Point 42°C to 45°C
Color Colorless
Flash Point >113°C (>235.4°F)
Packaging Glass Bottle
Quantity 5 g
UN Number NONH for all modes of transport
Beilstein 1619616
Formula Weight 264.32g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material 18-Crown-6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.