missing translation for 'onlineSavingsMsg'
Learn More

18-Crown-6, 99%

Product Code. 10226050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10226050 25 g Glass bottle 25g
10246000 5 g Glass bottle 5g
10134250 100 g Glass bottle 100g
11381110 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10226050 Supplier Thermo Scientific Acros Supplier No. 181560250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 17455-13-9 | C12H24O6 | 264.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17455-13-9
Molecular Formula C12H24O6
Molecular Weight (g/mol) 264.32
MDL Number MFCD00005113
InChI Key XEZNGIUYQVAUSS-UHFFFAOYSA-N
Synonym 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane
PubChem CID 28557
ChEBI CHEBI:32397
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCO1

Specifications

Melting Point 36.0°C to 40.0°C
Color Colorless or White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Fieser 04,142; 05,152; 06,133; 07,76; 08,1280; 09,126; 12,403; 15,76; 17,290
Merck Index 14,2602
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hydrocarbons,benzene,chloroform,methanol and trichlorotrifluoroethane
Formula Weight 264.32
Percent Purity 99%
Physical Form Crystalline Mass, Crystals or Liquid
Chemical Name or Material 18-Crown-6
Show More Show Less
compliance-icons
Product Identifier
  • 18-Crown-6
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.