missing translation for 'onlineSavingsMsg'
Learn More

18-Crown-6, TRC

Product Code. 30359659
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359659 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359659 Supplier Toronto Research Chemicals Supplier No. TRCC82487525G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 18-Crown-6
CAS 17455-13-9
Purity Grade Notes HPLC
Molecular Formula C12H24O6
Quantity 25 g
Synonym 18-Crown-6 ether, Ethylene Oxide Cyclic Hexamer, 1,4,7,10,13,16-Hexaoxacyclooctadecane, NSC 159836
InChI Formula InChI=1S/C12H24O6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2
SMILES C1COCCOCCOCCOCCOCCO1
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadecane
Molecular Weight (g/mol) 264.32
Formula Weight 264.1573
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.