missing translation for 'onlineSavingsMsg'
Learn More

18α-Glycyrrhetinic acid, MedChemExpress

Product Code. 16449066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449066 100 mg 100mg
16432376 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449066 Supplier Medchem Express Supplier No. HYN0375
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 18α-Glycyrrhetinic acid, a diet-derived compound, is an inhibitor of NF-kB and an activator of proteasome, which serves as pro-longevity and anti-aggregation factor in a multicellular organism. 18α-Glycyrrhetinic acid induces apoptosis.

18α-Glycyrrhetinic acid, a diet-derived compound, is an inhibitor of NF-kB and an activator of proteasome, which serves as pro-longevity and anti-aggregation factor in a multicellular organism. 18α-Glycyrrhetinic acid induces apoptosis.

Specifications

Chemical Name or Material 18α-Glycyrrhetinic acid
CAS 1449-05-4
Color White
Purity Grade Notes Research
Molecular Formula C30H46O4
Quantity 100 mg
Solubility Information DMSO : 19.23 mg/mL (40.86 mM; Need ultrasonic)
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@@]1([H])CC[C@]([C@@](CC[C@]3(C)[C@]4([H])C[C@@](C)(C(O)=O)CC3)(C)C4=C5)(C)[C@]2([H])C5=O
Molecular Weight (g/mol) 470.68
Formula Weight 470.68
Percent Purity 95.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.