missing translation for 'onlineSavingsMsg'
Learn More

18-O-Desmethyl Vinorelbine, TRC

Product Code. 30357184
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357184 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357184 Supplier Toronto Research Chemicals Supplier No. TRCD2965501MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 18-O-Desmethyl Vinorelbine
CAS 868596-47-8
Molecular Formula C44H52N4O8
Quantity 1 mg
Synonym (2beta,3beta,4beta,5alpha,12R,19alpha)-4-(Acetyloxy)-6,7-didehydro-15-[(2R,6R,8S)-8-carboxy-4-ethyl-1,3,6,7,8,9-hexahydro-2,6-methano-2H-azecino[4,3-b]indol-8-yl]-3-hydroxy-16-methoxy-1-methylaspidospermidine-3-carboxylic Acid Methyl Ester, Vinorelbine Impurity
InChI Formula InChI=1S/C44H52N4O8/c1-7-26-18-27-21-43(39(50)51,35-29(24-47(22-26)23-27)28-12-9-10-13-32(28)45-35)31-19-30-33(20-34(31)54-5)46(4)37-42(30)15-17-48-16-11-14-41(8-2,36(42)48)38(56-25(3)49)44(37,53)40(52)55-6/h9-14,18-20,27,36-38,45,53H,7-8,15-17,21-24H2,1-6H3,(H,50,51)/t27-,36-,37+,38+,41+,42+,43-,44-/m0/s1
SMILES CCC1=C[C@@H]2CN(C1)Cc3c([nH]c4ccccc34)[C@@](C2)(C(=O)O)c5cc6c(cc5OC)N(C)[C@H]7[C@](O)([C@H](OC(=O)C)[C@]8(CC)C=CCN9CC[C@]67[C@H]89)C(=O)OC
Molecular Weight (g/mol) 764.91
Formula Weight 764.3785
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.