missing translation for 'onlineSavingsMsg'
Learn More

18-Pentatriacontanone, tech. 85%

Product Code. 11374168
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374168 100 g 100g
11364168 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374168 Supplier Thermo Scientific Alfa Aesar Supplier No. 041445.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antiblocking agent | CAS: 504-53-0 | C35H70O | 506.94 g/mol

18-Pentatriacontanone is used as ant blocking agent. It is also used as pharmaceutical intermediate and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-53-0
Molecular Formula C35H70O
Molecular Weight (g/mol) 506.94
MDL Number MFCD00048491
InChI Key DMCJFWXGXUEHFD-UHFFFAOYSA-N
Synonym 18-pentatriacontanone, stearone, heptadecyl ketone, diheptadecyl ketone, di-n-heptadecyl ketone, unii-7in8ga6m68, stearon, stearyl ketone, di-stearyl ketone, acmc-1asrl
PubChem CID 10440
IUPAC Name pentatriacontan-18-one
SMILES CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 79°C to 80°C
Color White
Linear Formula (CH3(CH2)16)2CO
Quantity 100 g
Solubility Information Soluble in benzene
Formula Weight 506.94
Percent Purity 85%
Physical Form Powder
Chemical Name or Material 18-Pentatriacontanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.