missing translation for 'onlineSavingsMsg'
Learn More

18-Pentatriacontanone, TRC

Product Code. 30408978
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408978 Supplier Toronto Research Chemicals Supplier No. TRCP5045301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 18-Pentatriacontanone
CAS 504-53-0
Molecular Formula C35H70O
Quantity 1 g
Synonym Di-n-heptadecyl Ketone, EP 2036-18, Heptadecyl Ketone, NSC 83612, Stearone, Wax KS
InChI Formula InChI=1S/C35H70O/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(36)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-34H2,1-2H3
SMILES CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC
IUPAC Name pentatriacontan-18-one
Molecular Weight (g/mol) 506.93
Formula Weight 506.5427
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.