missing translation for 'onlineSavingsMsg'
Learn More

18Alpha-Glycyrrhetinic Acid, TRC

Product Code. 30402967
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402967 100 mg 100mg
30391758 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402967 Supplier Toronto Research Chemicals Supplier No. TRCG735005100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 18alpha-Glycyrrhetinic acid
CAS 1449-05-4
Molecular Formula C30 H46 O4
Quantity 100 mg
Synonym Olean-12-en-29-oic acid, 3-hydroxy-11-oxo-, (3β,18α,20β)-, 18α-Olean-12-en-30-oic acid, 3β-hydroxy-11-oxo- (6CI,7CI,8CI), (3β,18α,20β)-3-Hydroxy-11-oxoolean-12-en-29-oic acid, 18-Isoglycyrrhetinic acid, 18α-Glycyrrhetic acid, 18α-Glycyrrhetinic acid, NSC 35350, STX 353, β-Glycyrrhetinic acid
InChI Formula InChI=1S/C30H46O4/c1-25(2)21-8-11-30(7)23(28(21,5)10-9-22(25)32)20(31)16-18-19-17-27(4,24(33)34)13-12-26(19,3)14-15-29(18,30)6/h16,19,21-23,32H,8-15,17H2,1-7H3,(H,33,34)/t19-,21+,22+,23-,26-,27+,28+,29-,30-/m1/s1
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(=O)O
IUPAC Name (2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bS)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
Molecular Weight (g/mol) 470.68
Formula Weight 470.3396
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.