missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 18mm Magnetic Universal Screw Seal, Silver, Center Hole, Precision Thread, Assembled Septum

Product Code. 11895000 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue Transparent, White, Silver
Blue, White, Silver
Gray, Red, Silver
Red, White, Silver
Silver, White
Material (Liner):
PTFE/Butyl
Silicone/Aluminum
Silicone/PTFE
Thickness (Metric):
1.3 mm
1.5 mm
1.6 mm
Unit Size:
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material (Liner) Thickness (Metric) unitSize
11895000 Silver, White Silicone/Aluminum 1.3 mm Pack of 1000
11787607 Gray, Red, Silver PTFE/Butyl 1.6 mm Pack of 1000
11778256 Red, White, Silver Silicone/PTFE 1.3 mm Pack of 1000
11865000 Blue Transparent, White, Silver Silicone/PTFE 1.3 mm Pack of 1000
11853912 Blue, White, Silver Silicone/PTFE 1.5 mm Pack of 1000
11713027 Red, White, Silver Silicone/PTFE 1.5 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11895000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pre-assembled caps and septa are convenient and minimize contamination from handling.

  • 18mm
  • Magnetic
  • Universal Screw Seal
  • Silver
  • Center hole
  • Precision Thread
  • Assembled septum
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Silicone/Aluminum
Color Silver, White
Diameter (Metric) Outer 18 mm
Material Steel
For Use With (Application) Headspace
Quantity 1000
Thickness (Metric) 1.3 mm
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.