Learn More
Abnova™ 19p13.3/19q13.13 FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0159.200uL
Additional Details : Weight : 0.02000kg
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
Liquid |
FISH-Ce | |
200 μL |