missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Propylene-d6 Oxide, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Propylene oxide-d6 |
| CAS | 202468-69-7 |
| Molecular Formula | C3 2H6 O |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/i1D3,2D2,3 D |
| SMILES | [2 H]C([2 H])([2 H])C1([2 H])OC1([2 H])[2 H] |
| IUPAC Name | 2,2,3-trideuterio-3-(trideuteriomethyl)oxirane |
| Molecular Weight (g/mol) | 64.12 |
| Formula Weight | 64.0795 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction