missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dichloro-iso-propyl-d5 Alcohol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,3-Dichloropropan-2-ol-d5 |
| CAS | 1173020-20-6 |
| Molecular Formula | C3 D5 H Cl2 O |
| Quantity | 1 g |
| Synonym | d5-1,3-Dichloro-2-propanol, 1,3-Dichloro-2-propanol-d5, 1,3-Dichloro-2-hydroxypropane-d5, 1,3-Dichlorohydrin-d5, 1,3-Dichloroisopropanol-d5, 1,3-Dichloroisopropyl Alcohol-d5, 2-Chloro-1-(chloromethyl)ethanol-d5, Bis(chloromethyl)methanol-d5, Glycerol 1,3-Dichlorohydrin-d5, NSC 70982, Propylene Dichlorohydrin |
| InChI Formula | InChI=1 S/C3H6Cl2O/c4-1-3(6)2-5/h3,6 H,1-2H2/i1D2,2D2,3 D |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])(O)C([2 H])([2 H])Cl |
| IUPAC Name | 1,3-dichloro-1,1,2,3,3-pentadeuteriopropan-2-ol |
| Molecular Weight (g/mol) | 134.02 |
| Formula Weight | 133.011 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction