missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylpyridine-d9, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dimethylpyridine-D9 |
| CAS | 350818-65-4 |
| Molecular Formula | C7 D9 N |
| Quantity | 1 g |
| Synonym | Pyridine-2,3,4-d3, 5,6-di(methyl-d3)- (9 CI), 5,6-Di(methyl-d3)pyridine-2,3,4-d3, 2,3-Dimethylpyridine-d9, 2,3-Dimethylpyridine-D9, 2,3-Lutidine-d9 |
| InChI Formula | InChI=1 S/C7H9N/c1-6-4-3-5-8-7(6)2/h3-5 H,1-2H3/i1D3,2D3,3 D,4 D,5 D |
| SMILES | [2 H]c1nc(c(c([2 H])c1[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4-trideuterio-5,6-bis(trideuteriomethyl)pyridine |
| Molecular Weight (g/mol) | 116.2085 |
| Formula Weight | 116.13 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction