missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromotoluene-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromotoluene-3,4,5,6-D4 |
| CAS | 56444-57-6 |
| Molecular Formula | C7 D4 H3 Br |
| Quantity | 1 g |
| Synonym | Benzene-1,2,3,4-d4, 5-bromo-6-methyl- (9 CI, ACI), 5-Bromo-6-methylbenzene-1,2,3,4-d4 (ACI), 2-Bromotoluene-d4, 2-Bromotoluene-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C7H7Br/c1-6-4-2-3-5-7(6)8/h2-5 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Br)c(C)c1[2 H] |
| IUPAC Name | 1-bromo-2,3,4,5-tetradeuterio-6-methylbenzene |
| Molecular Weight (g/mol) | 175.0591 |
| Formula Weight | 173.9982 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction