missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropropane-1,1,1,3,3,3-d6, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 2-Chloropropane-1,1,1,3,3,3-D6 |
| CAS | 23197-02-6 |
| Molecular Formula | C3 D6 H Cl |
| Quantity | 1 g |
| Synonym | Propane-1,1,1,3,3,3-d6, 2-chloro- (8 CI, 9 CI), 1,1,1,3,3,3-Hexadeutero-2-chloropropane, 2-Chloro-1,1,1,3,3,3-hexadeuteropropane, 2-Chloropropane-1,1,1,3,3,3-D6, 2-Chloropropane-1,1,1,3,3,3-d6, 2-Chloropropane-d6 |
| InChI Formula | InChI=1 S/C3H7Cl/c1-3(2)4/h3H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Cl)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-chloro-1,1,1,3,3,3-hexadeuteriopropane |
| Molecular Weight (g/mol) | 84.5777 |
| Formula Weight | 84.0613 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction