missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-d3-phenol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyphenol-d3 |
| CAS | 74495-69-5 |
| Molecular Formula | C7 2H3 H5 O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccccc1O |
| IUPAC Name | 2-(trideuteriomethoxy)phenol |
| Molecular Weight (g/mol) | 127.16 |
| Formula Weight | 127.0713 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction