missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-d3-propionic Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-propanoic-3,3,3-d3 Acid |
| CAS | 95926-99-1 |
| Molecular Formula | C4 D3 H5 O2 |
| Quantity | 1 g |
| Synonym | Propanoic-3,3,3-d3 acid, 2-methyl- (9 CI), 2-Methylpropanoic-3,3,3-d3 acid, Isobutyric-3,3,3-d6 acid, 2-Methylpropanoic Acid-3,3,3-D3, Isobutyric acid D3 (methyl-D3), Isobutyric acid-d3, 2-Methyl-d3-propionic Acid |
| InChI Formula | InChI=1 S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C)C(=O)O |
| IUPAC Name | 3,3,3-trideuterio-2-methylpropanoic acid |
| Molecular Weight (g/mol) | 91.124 |
| Formula Weight | 91.071 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction