missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)phenol-2,4,6-d3,OD, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 3-(Trifluoromethyl)phenol-O,2,4,6-D4 |
| CAS | 1219798-47-6 |
| Molecular Formula | C7 D4 H F3 O |
| Quantity | 1 g |
| Synonym | 3-(Trifluoromethyl)phenol-2,4,6-d3,OD, 3-(Trifluoromethyl)phenol-d4, 3-(Trifluoromethyl)phenol-O,2,4,6-D4, α,α,α-Trifluoro-m-cresol-d4 |
| InChI Formula | InChI=1 S/C7H5F3O/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4,11 H/i2D,3 D,4 D/hD |
| SMILES | [2 H]Oc1c([2 H])cc([2 H])c(c1[2 H])C(F)(F)F |
| IUPAC Name | 1,3,5-trideuterio-2-deuteriooxy-4-(trifluoromethyl)benzene |
| Molecular Weight (g/mol) | 166.1339 |
| Formula Weight | 166.0544 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction