missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminophenol-d7, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
1 g
missing translation for 'unitSize'
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Aminophenol-d7 |
| CAS | 285132-88-9 |
| Molecular Formula | C6 2H7 N O |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8 H,7H2/i1D,2 D,3 D,4 D/hD3 |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(N([2 H])[2 H])c([2 H])c1[2 H] |
| IUPAC Name | N,N,2,3,5,6-hexadeuterio-4-deuteriooxyaniline |
| Molecular Weight (g/mol) | 116.17 |
| Formula Weight | 116.0967 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction