missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Lysine-2-15N 2HCl, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | DL-Lysine-2-15 N Dihydrochloride |
| CAS | 2747-89-9 |
| Molecular Formula | C6 H14 15 N N O2 . 2 Cl H |
| Quantity | 1 g |
| Synonym | Lysine-2-15 N, hydrochloride (1:2) (ACI), DL-Lysine-2-15 N, dihydrochloride (ZCI), Lysine-2-15 N, dihydrochloride (9 CI), DL-Lysine-2-15 N Dihydrochloride, DL-Lysine-N2-15 N hydrochloride (1:2), DL-Lysine-2-15 N 2 HCl, DL-Lysine-2-15 N 2 HCl, DL-Lysine-2-15 N 2 HCl, DL-Lysine-2-15 N 2 HCl |
| InChI Formula | InChI=1 S/C6H14N2O2.2 ClH/c7-4-2-1-3-5(8)6(9)10;;/h5H,1-4,7-8H2,(H,9,10);2*1 H/i8+1;; |
| SMILES | Cl.Cl.[15 NH2]C(CCCCN)C(=O)O |
| IUPAC Name | 6-amino-2-(15 N)azanylhexanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 220.1029 |
| Formula Weight | 219.0559 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction