missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-5-(2-Bromovinyl)uracil, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (E)-5-(2-Bromovinyl)uracil |
| CAS | 69304-49-0 |
| Molecular Formula | C6 H5 Br N2 O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H5BrN2O2/c7-2-1-4-3-8-6(11)9-5(4)10/h1-3H,(H2,8,9,10,11)/b2-1+ |
| SMILES | Br\C=C\C1=CNC(=O)NC1=O |
| IUPAC Name | 5-[(E)-2-bromoethenyl]-1H-pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 217.02 |
| Formula Weight | 215.9534 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction