missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Cresol-d8, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | p-Cresol-d8 |
| CAS | 190780-66-6 |
| Molecular Formula | C7 2H8 O |
| Quantity | 1 g |
| Synonym | p-Cresol (D8) |
| InChI Formula | InChI=1 S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8 H,1H3/i1D3,2 D,3 D,4 D,5 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-deuteriooxy-6-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 116.19 |
| Formula Weight | 116.1077 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction