missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Terphenyl-d14, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | p-Terphenyl D14 |
| CAS | 1718-51-0 |
| Molecular Formula | C18 2H14 |
| Quantity | 1 g |
| Synonym | 1,1':4',1''-Terphenyl-2,2',2'',3,3',3'',4,4'',5,5',5'',6,6',6''-d14, p-Terphenyl-d14 (7 CI,8 CI), 4-Terphenyl-d14, p-Terphenyl D14 |
| InChI Formula | InChI=1 S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c(c([2 H])c2[2 H])c3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene |
| Molecular Weight (g/mol) | 244.39 |
| Formula Weight | 244.1974 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction