missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Phenol-d5 |
| CAS | 4165-62-2 |
| Molecular Formula | C6 D5 H O |
| Quantity | 1 g |
| Synonym | Phenol-2,3,4,5,6 D5, Phenol (ring-D5), Phen-2,3,4,5,6-d5-ol, Phen-d5-ol (7 CI,8 CI,9 CI), 2,3,4,5,6-Pentadeuteriophenol |
| InChI Formula | InChI=1 S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(O)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-Pentadeuteriophenol |
| Molecular Weight (g/mol) | 99.14 |
| Formula Weight | 99.0732 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction