missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Cinnamic-d7 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | (2 E)-3-(Phenyl-2,3,4,5,6-d5)-2-propenoic-2,3-d2 Acid |
| CAS | 343338-31-8 |
| Molecular Formula | C9 D7 H O2 |
| Quantity | 1 g |
| Synonym | 2-Propenoic-2,3-d2 acid, 3-(phenyl-2,3,4,5,6-d5)-, (2 E)- (ACI), (2 E)-3-(Phenyl-2,3,4,5,6-d5)-2-propenoic-2,3-d2 acid (ACI), 2-Propenoic-2,3-d2 acid, 3-(phenyl-d5)-, (2 E)- (9 CI), trans-Cinnamic-d7 acid, trans-Cinnamic acid D7 |
| InChI Formula | InChI=1 S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7 H,(H,10,11)/b7-6+/i1D,2 D,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]\C(=C(\[2 H])/c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H])\C(=O)O |
| IUPAC Name | (E)-2,3-dideuterio-3-(2,3,4,5,6-pentadeuteriophenyl)prop-2-enoic acid |
| Molecular Weight (g/mol) | 155.2 |
| Formula Weight | 155.0964 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction