missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,3-Benzotriazol-5-amine, TRC

Product Code. 30329917
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329917 Supplier Toronto Research Chemicals Supplier No. TRCB5405405G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-1,2,3-Benzotriazol-5-amine
CAS 3325-11-9
Molecular Formula C6 H6 N4
Quantity 5 g
Synonym 1H-Benzotriazol-5-amine, (1H-Benzo[d][1,2,3]triazol-6-yl)amine, 1H-1,2,3-Benzotriazol-6-amine, 1H-Benzo[d][1,2,3]triazol-5-amine, 1H-Benzo[d][1,2,3]triazol-6-amine, 5(6)-Aminobenzotriazole, 5-Aminobenzotriazole, 5-Aminobenztriazole, 6-Aminobenzotriazole
InChI Formula InChI=1S/C6H6N4/c7-4-1-2-5-6(3-4)9-10-8-5/h1-3H,7H2,(H,8,9,10)
SMILES Nc1ccc2n[nH]nc2c1
IUPAC Name 2H-benzotriazol-5-amine
Molecular Weight (g/mol) 134.139
Formula Weight 134.059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.