missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%

Product Code. 11461067
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461067 25 g 25g
11451067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461067 Supplier Thermo Scientific Alfa Aesar Supplier No. B24075.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 2043-55-2 | C6H4F9I | 373.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-55-2
Molecular Formula C6H4F9I
Molecular Weight (g/mol) 373.988
MDL Number MFCD00039409
InChI Key CXHFIVFPHDGZIS-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane
PubChem CID 74887
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
SMILES C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point -25°C
Density 1.94
Boiling Point 138°C to 140°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.37
Quantity 25 g
Beilstein 1870862
Sensitivity Light sensitive
Formula Weight 373.99
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iodohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.