missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™

Product Code. 11349255
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349255 25 g 25g
11329255 1 g 1g
11339255 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349255 Supplier Thermo Scientific Alfa Aesar Supplier No. L16585.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.

Solubility
Miscible with ethanol and tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101947-16-4
Molecular Formula C16H19F17O3Si
Molecular Weight (g/mol) 610.38
MDL Number MFCD00042334
InChI Key MLXDKRSDUJLNAB-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g
PubChem CID 145374
IUPAC Name triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
SMILES CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC

Specifications

Boiling Point 103°C to 106°C (3mmHg)
Quantity 25 g
Linear Formula CF3(CF2)7(CH2)2Si(OCH2CH3)3
UN Number UN1760
Flash Point 100°C (212°F)
Solubility Information Miscible with ethanol and tetrahydrofuran.
Formula Weight 610.38
Percent Purity 97%
Refractive Index 1.342
Sensitivity Moisture sensitive
Density 1.41
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyltriethoxysilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.