missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorododecan-1-ol, TRC

Product Code. 30426396
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426396 Supplier Toronto Research Chemicals Supplier No. TRCP2862251G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-dodecanol
CAS 865-86-1
Molecular Formula C12 H5 F21 O
Quantity 1 g
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecanol, 1,1,2,2-Tetrahydrohenicosafluorododecanol, 1,1,2,2-Tetrahydroperfluorododecanol, 10:2 FTOH, 10:2 Fluorotelomer alcohol, 1H,1H,2H,2H-Perfluorododecan-1-ol, 1H,1H,2H,2H-Perfluorododecanol, 2-(Perfluorodecyl)ethanol, 2-(Perfluorodecyl)ethyl alcohol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12','12,12-Heneicosafluorododecan-1-ol, 3-(Perfluorodecyl)ethanol, A 2020, A 2020 (fluoro compound), PFA 10
InChI Formula InChI=1S/C12H5F21O/c13-3(14,1-2-34)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)11(29,30)12(31,32)33/h34H,1-2H2
SMILES OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
Molecular Weight (g/mol) 564.13
Formula Weight 564.0005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.