missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorohexyl iodide, TRC

Product Code. 30418097
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418097 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418097 Supplier Toronto Research Chemicals Supplier No. TRCP28637010G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H,1H,2H,2H-Perfluorohexyl iodide
CAS 2043-55-2
Purity Grade Notes GC
Molecular Formula C6 H4 F9 I
Quantity 10 g
Synonym Hexane','1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo- (7CI','8CI','9CI','ACI), 1,1,1,2,2,3,3,4,4-Nonafluoro-6-iodohexane (ACI), 1-Iodo-1H,1H,2H,2H-perfluorohexane, 1-Iodo-2-(perfluorobutyl)ethane, 1-Iodo-3,3,4,4,5,5,6,6,6-nonafluorohexane, 1-Perfluorobutyl-2-iodoethane, 1H,1H,2H,2H-Nonafluorohexyl iodide, 1H,1H,2H,2H-Perfluorohexyl iodide, 2-(Nonafluorobutyl)ethyl iodide, 2-(Perfluorobutyl)ethyl iodide, 2-Perfluorobutyl-1-iodoethane, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl iodide
InChI Formula InChI=1S/C6H4F9I/c7-3(8,1-2-16)4(9,10)5(11,12)6(13,14)15/h1-2H2
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
Molecular Weight (g/mol) 373.986
Formula Weight 373.921
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.