missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctanol, 97%

Product Code. 11490701
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490701 50 g 50g
11480701 10 g 10g
11400711 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490701 Supplier Thermo Scientific Alfa Aesar Supplier No. B20156.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 647-42-7 | C8H5F13O | 364.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctanol is used to prepare fluorinated aromatic diamine by reacting with 2-(4-nitrophenoxy)-4-nitrophenol through Mitsunobu reaction, which one reacts with dianhydrides such as 4,4′-(hexafluoroisopropylidene)diphthalic anhydride (6FDA), pyromellitic dianhydride (PMDA) and benzophenonetetracarboxylic dianhydride (BTDA) to prepare polyimides.

Solubility
Miscible with chloroform and methanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 647-42-7
Molecular Formula C8H5F13O
Molecular Weight (g/mol) 364.106
MDL Number MFCD00042143
InChI Key GRJRKPMIRMSBNK-UHFFFAOYSA-N
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol
PubChem CID 69537
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
SMILES C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.68
Boiling Point 90°C (20 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.314
Quantity 50 g
Beilstein 1992757
Solubility Information Miscible with chloroform and methanol. Immiscible with water.
Formula Weight 364.11
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.