missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctyl-1-thiol, TRC

Product Code. 30421267
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421267 Supplier Toronto Research Chemicals Supplier No. TRCP2861251G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyl-1-thiol
CAS 34451-26-8
Purity Grade Notes GC
Molecular Formula C8H5F13S
Quantity 1 g
Synonym 1-Octanethiol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro- (9CI','ACI): 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol (ACI), 1,1,2,2-Tetrahydroperfluorooctanethiol, 1,1,2,2-Tetrahydroperfluorooctyl mercaptan, 1H,1H,1H,2H-Perfluorooctanethiol, 1H,1H,2H,2H-Perfluorooctanethiol, 2-(Perfluorohexyl)ethane-1-thiol, 2-(Perfluorohexyl)ethanethiol, Foralkyl EM 6
InChI Formula InChI=1S/C8H5F13S/c9-3(10,1-2-22)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h22H,1-2H2
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCS
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctane-1-thiol
Molecular Weight (g/mol) 380.17
Formula Weight 379.9904
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.