missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane, 97%

Product Code. 11309285
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309285 5 g 5g
11319285 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309285 Supplier Thermo Scientific Alfa Aesar Supplier No. L16605.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73609-36-6 | C9H7Cl2F13Si | 461.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73609-36-6
Molecular Formula C9H7Cl2F13Si
Molecular Weight (g/mol) 461.12
MDL Number MFCD00042360
InChI Key VBDMVWQNRXVEGC-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyl methyldichlorosilane, 1h,1h,2h,2h-perfluorooctylmethyldichlorosilane, dichloro methyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl methyldichlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-methyldichlorosilane, dichloro-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, bis chloranyl-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecakis fluoranyl octyl silane, silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, 2-tridecafluorohexyl ethyldichloro methyl silane
PubChem CID 123573
IUPAC Name dichloro-methyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C[Si](Cl)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Boiling Point 189°C to 190°C
Quantity 5 g
Flash Point 51°C (123°F)
Refractive Index 1.35
Sensitivity Moisture sensitive
Density 1.55
UN Number UN2920
Beilstein 6153667
Formula Weight 461.13
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane
Show More Show Less

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.