missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-dodecene, 97%

Product Code. 11349265
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349265 5 g 5g
11359265 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349265 Supplier Thermo Scientific Alfa Aesar Supplier No. L16591.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 30389-25-4 | C12H3F21 | 546.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30389-25-4
Molecular Formula C12H3F21
Molecular Weight (g/mol) 546.122
MDL Number MFCD00042346
InChI Key UCHSAVGOZUCXHC-UHFFFAOYSA-N
Synonym perfluorodecyl ethylene, 1h,1h,2h-perfluoro-1-dodecene, acmc-20aplc, n-perfluorodecyl ethylene, heneicosafluorodecyl-ethylene, 1h,1h,2h-perfluorododec-1-ene, heneicosafluorodecyl ethylene
PubChem CID 121692
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododec-1-ene
SMILES C=CC(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.711
Boiling Point 71°C to 72°C (14 mmHg)
Linear Formula CF3(CF2)9CH=CH2
Refractive Index 1.305
Quantity 5 g
Beilstein 2493721
Formula Weight 546.12
Percent Purity 97%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-dodecene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.