missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-decanol, 98%

Product Code. 11319245
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319245 5 g 5g
11329245 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319245 Supplier Thermo Scientific Alfa Aesar Supplier No. L16580.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 307-37-9 | C10H3F19O | 500.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-37-9
Molecular Formula C10H3F19O
Molecular Weight (g/mol) 500.103
MDL Number MFCD00153234
InChI Key NIRPXSQCRWXHNZ-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-decanol, 1h,1h-nonadecafluoro-1-decanol, 1h,1h-perfluorodecan-1-ol, nonadecafluorononyl methanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decan-1-ol
PubChem CID 2733275
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecan-1-ol
SMILES C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 83°C to 90°C
Boiling Point 95°C (20 mmHg)
Linear Formula CF3(CF2)8CH2OH
Quantity 5 g
Beilstein 1810012
Formula Weight 500.1
Percent Purity 98%
Chemical Name or Material 1H,1H-Perfluoro-1-decanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.