missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-dodecanol, tech. 90%

Product Code. 11339265
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339265 25 g 25g
11329265 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339265 Supplier Thermo Scientific Alfa Aesar Supplier No. L16590.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 423-65-4 | C12H3F23O | 600.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-65-4
Molecular Formula C12H3F23O
Molecular Weight (g/mol) 600.12
MDL Number MFCD00153235
InChI Key SHTZQFTXUMCALC-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-dodecanol, 1h,1h-perfluorododecan-1-ol, 1h,1h-perfluorododecanol, tricosafluoroundecylmethanol, 1h,1h-tricosafluoro-1-dodecanol, 1h,1h-perfluoro-1-lauryl alcohol
PubChem CID 2760321
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point ∼110°C
Boiling Point 224°C (740 mmHg)
Quantity 25 g
Beilstein 1810966
Formula Weight 600.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 1H,1H-Perfluoro-1-dodecanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.