missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-2-carboxylic acid hydrate, 90%, Thermo Scientific™

Product Code. 10096202
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10096202 1 g Amber Glass Bottle 1g
10187472 250 mg Amber Glass Bottle 250mg
10230545 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096202 Supplier Thermo Scientific Maybridge Supplier No. CC09051DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 849776-47-2
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
InChI Key YQHUUYLPOCNBKD-UHFFFAOYSA-N
Synonym 1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1
PubChem CID 18525703
IUPAC Name 1H-benzimidazole-2-carboxylic acid;hydrate
SMILES C1=CC=C2C(=C1)NC(=N2)C(=O)O.O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.17
Percent Purity 90%
Chemical Name or Material 1H-Benzimidazole-2-carboxylic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.