missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10794255
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10794255 250 mg Amber Glass Bottle 250mg
10092314 1 g Amber Glass Bottle 1g
10055483 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10794255 Supplier Thermo Scientific Maybridge Supplier No. CC45904CB
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58442-17-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
InChI Key ALCHVVTYAHQOFY-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866
PubChem CID 10012038
IUPAC Name 3H-benzimidazole-5-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)NC=N2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 1H-Benzimidazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.