missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98%

Product Code. 11463187
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463187 25 g 25g
11443187 1 g 1g
11453187 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463187 Supplier Thermo Scientific Alfa Aesar Supplier No. A16140.14
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56602-33-6 | C12H22F6N6OP2 | 442.287 g/mol

1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

Solubility
Soluble in methanol, acetone and dichloromethane. Insoluble in water.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56602-33-6
Molecular Formula C12H22F6N6OP2
Molecular Weight (g/mol) 442.287
MDL Number MFCD00011948
InChI Key MGEVGECQZUIPSV-UHFFFAOYSA-N
Synonym bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
PubChem CID 151348
IUPAC Name benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
SMILES CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∼138°C (decomposition)
Quantity 25 g
UN Number UN1325
Beilstein 4287025
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol,acetone and dichloromethane. Insoluble in water.
Formula Weight 442.29
Percent Purity 98%
Chemical Name or Material 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.