missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole, ≥99.0%, Solstice

Product Code. 15681190
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15681190 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15681190 Supplier Honeywell-Fluka Supplier No. 12798100G
  • ONLINE EXCLUSIVE PRICE
    99.12 €
    93.26 € / 100g
    Save 5.86 €
    5% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for amperometric det. of Cu,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 95-14-7
Melting Point 95°C to 99°C
Color Beige
pH 6.0 to 7.0 (at 100g/L 20°C)
Flash Point 170°C (338°F)
Ignition Residue ≤0.1%
Packaging Poly bottle
Molecular Formula C6H5N3
MDL Number MFCD00005699
Quantity 100 g
UN Number NONH for all modes of transport
Beilstein 112133
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
SMILES C1=CC2=NNN=C2C=C1
IUPAC Name 2H-benzotriazole
Molecular Weight (g/mol) 119.127
PubChem CID 7220
ChEBI CHEBI:75331
Formula Weight 119.12g/mol
Percent Purity ≥99.0%
Physical Form Solid
Chemical Name or Material 1H-Benzotriazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.