missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazol-2-ylmethanol hydrochloride, 97%, Thermo Scientific™

Product Code. 10452434
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10452434 5 g Amber Glass Bottle 5g
10160632 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10452434 Supplier Thermo Scientific Maybridge Supplier No. CC08909DE
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 116177-22-1
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.563
MDL Number MFCD03659700
InChI Key FEORNCWZOSTSRO-UHFFFAOYSA-N
Synonym 1h-imidazol-2-ylmethanol hydrochloride, 1h-imidazol-2-yl methanol hydrochloride, imidazolylmethanol hydrochloride, hydroxymethylimidazole hydrochloride, hydroxymethyl imidazole hydrochloride, imidazol-2-ylmethan-1-ol, chloride
PubChem CID 2776278
IUPAC Name 1H-imidazol-2-ylmethanol;hydrochloride
SMILES C1=CN=C(N1)CO.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 134.57
Percent Purity 97%
Chemical Name or Material 1H-imidazol-2-ylmethanol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.