missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-2-carboxaldehyde, TRC

Product Code. 30393529
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393529 5 g 5g
30395978 2.5 g 2.5g
30408766 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393529 Supplier Toronto Research Chemicals Supplier No. TRCI3502155G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10111-08-7
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
Synonym Imidazole-2-carboxaldehyde, 1H-Imidazole-2-carbaldehyde, 2-Formylimidazole, 2-Imidazolecarbaldehyde, 2-Imidazolylformaldehyde
IUPAC Name 1H-imidazole-2-carbaldehyde
SMILES O=Cc1ncc[nH]1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H4N2O/c7-3-4-5-1-2-6-4/h1-3H,(H,5,6)
Formula Weight 96.0324
Chemical Name or Material 1H-Imidazole-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.