missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10228262
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10228262 1 g Amber Glass Bottle 1g
10081134 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10228262 Supplier Thermo Scientific Maybridge Supplier No. CC08901DA
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 16042-25-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
InChI Key KYWMCFOWDYFYLV-UHFFFAOYSA-N
Synonym 2-imidazolecarboxylic acid, imidazole-2-carboxylic acid, unii-o9g6515vc1, 1h-imidazolecarboxylic acid, 2-imidazolecarboxylicacid, 2-imidazocarboxylic acid, 1h-imidazole-2-carboxylicacid, imidazolecarboxylic, acmc-1avrt, 2-carboxy-1h-imidazole
PubChem CID 574321
IUPAC Name 1H-imidazole-2-carboxylic acid
SMILES C1=CN=C(N1)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material 1H-Imidazole-2-carboxylic acid
compliance-icons
Product Identifier
  • 1H-Imidazole-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Imidazole-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.