missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10556884
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10556884 25 g Amber Glass Bottle 25g
10316654 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10556884 Supplier Thermo Scientific Maybridge Supplier No. CC08804EB
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3034-50-2
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00173726
InChI Key ZQEXIXXJFSQPNA-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carbaldehyde, imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, imidazole-4-carboxaldehyde, 4-imidazolecarboxaldehyde, 4-formylimidazole, 4 5-imidazolecarboxaldehyde, 3h-imidazole-4-carbaldehyde, 4-formyl-1h-imidazole, imidazole-5-carbaldehyde
PubChem CID 76428
IUPAC Name 1H-imidazole-5-carbaldehyde
SMILES C1=C(NC=N1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 96.09
Percent Purity 97%
Chemical Name or Material 1H-imidazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.