missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-4-carboxaldehyde, 97%

Product Code. 10220800 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10220800

Brand: Thermo Scientific Acros 431030050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 669050-70-8 | C8H6N2O | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 669050-70-8
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD06738279
InChI Key FPJXNCKSPFGQGC-UHFFFAOYSA-N
Synonym 4-formylindazole, 1h-indazole-4-carboxaldehyde, 4-formyl-1h-indazole, 1h-indazol-4-carboxaldehyde, pubchem22512, acmc-209nwx, 1h-indazole-4-carboxaldehyde 9ci
PubChem CID 45083569
IUPAC Name 1H-indazole-4-carbaldehyde
SMILES C1=CC(=C2C=NNC2=C1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 166.0°C to 167.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 146.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazole-4-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazole-4-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.